The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008387.2 | NP_001008388.1 | 5 - 6 | 661 | 140 |
NM_001270040.1 | NP_001256969.1 | 4 - 5 | 523 | 140 | |
NM_198448.3 | NP_940850.1 | 5 - 6 | 561 | 140 | |
XM_005264135.2 | XP_005264192.1 | 5 - 6 | 723 | 140 | |
GenBank mRNA | AB161037.1 | - | 5 - 6 | 546 | 140 |
AB161038.1 | - | 5 - 6 | 646 | 140 | |
AI025191.1 | - | 1 - 2 | 140 | ||
AK292595.1 | - | 5 - 6 | 545 | 140 | |
AY359047.1 | - | 5 - 6 | 547 | 140 | |
AY428734.1 | - | 5 - 6 | 520 | 140 | |
BC103852.2 | - | 4 - 5 | 482 | 140 | |
BC103853.1 | - | 5 - 6 | 620 | 140 | |
BC103854.1 | - | 5 - 6 | 520 | 140 | |
BI792387.1 | - | 1 - 2 | 140 |