The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004342.3 | NP_001004342.3 | 1 - 2 | 1089 | 168 |
NM_001300889.1 | NP_001287818.1 | 3 - 4 | 903 | 168 | |
XM_011544192.2 | XP_011542494.1 | 1 - 2 | 1089 | 168 | |
XM_017001323.1 | XP_016856812.1 | 1 - 2 | 1089 | 168 | |
GenBank mRNA | AK126782.1 | - | 1 - 2 | 1089 | 168 |
AY253917.1 | - | 3 - 4 | 861 | 168 |