The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173562.4 | NP_775833.2 | 2 - 3 | 551 | 108 |
XM_005248940.4 | XP_005248997.1 | 3 - 4 | 406 | 108 | |
XM_006715023.2 | XP_006715086.1 | 2 - 3 | 324 | 108 | |
XM_011514398.2 | XP_011512700.1 | 3 - 4 | 402 | 108 | |
GenBank mRNA | AK304377.1 | - | 2 - 3 | 272 | 108 |
AY305862.1 | - | 2 - 3 | 551 | 108 |