The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286579.1 | NP_001273508.1 | 4 - 5 | 858 | 144 |
NM_001286580.1 | NP_001273509.1 | 6 - 7 | 1082 | 144 | |
NM_173562.4 | NP_775833.2 | 7 - 8 | 1356 | 144 | |
NR_104480.1 | 5 - 6 | 961 | 144 | ||
NR_104481.1 | 3 - 4 | 652 | 144 | ||
XM_005248940.4 | XP_005248997.1 | 8 - 9 | 1211 | 144 | |
XM_006715023.2 | XP_006715086.1 | 7 - 8 | 1129 | 144 | |
XM_011514398.2 | XP_011512700.1 | 8 - 9 | 1207 | 144 | |
GenBank mRNA | AK295717.1 | - | 3 - 4 | 402 | 144 |
AK302861.1 | - | 6 - 7 | 813 | 144 | |
AK304377.1 | - | 7 - 8 | 1077 | 144 | |
AK316456.1 | - | 5 - 6 | 707 | 144 | |
AL832381.1 | - | 6 - 7 | 868 | 144 | |
AY305862.1 | - | 7 - 8 | 1356 | 144 | |
BC023525.2 | - | 4 - 5 | 596 | 144 |