Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLRA4
Entrez Gene ID
441509
Gene Name
glycine receptor alpha 4
Gene Aliases
-
Chromosome Location
Chr.X: 103707344 - 103728624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024452.2 NP_001019623.2 1 - 2 495 84
NM_001172285.1 NP_001165756.1 1 - 2 495 84
GenBank mRNA AK309010.1 - 1 - 2 495 84

View Full Product Details