The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284358.1 | NP_001271287.1 | 6 - 7 | 814 | 127 |
NM_001284361.1 | NP_001271290.1 | 9 - 10 | 1063 | 193 | |
NM_001284362.1 | NP_001271291.1 | 8 - 9 | 997 | 127 | |
NM_033030.5 | NP_149019.1 | 8 - 9 | 910 | 127 | |
NM_197970.2 | NP_932074.1 | 8 - 9 | 847 | 127 | |
XM_006712715.3 | XP_006712778.1 | 8 - 9 | 1207 | 127 | |
XM_011511692.2 | XP_011509994.1 | 9 - 10 | 1273 | 193 | |
XM_011511693.2 | XP_011509995.1 | 9 - 10 | 1273 | 193 | |
XM_011511694.2 | XP_011509996.1 | 9 - 10 | 880 | 193 | |
GenBank mRNA | AF272858.1 | - | 8 - 9 | 855 | 127 |
AK058099.1 | - | 8 - 9 | 910 | 127 | |
AK093453.1 | - | 9 - 10 | 1063 | 193 | |
AK097795.1 | - | 8 - 9 | 1206 | 127 | |
AK098824.1 | - | 8 - 9 | 879 | 127 | |
AK302817.1 | - | 6 - 7 | 814 | 127 | |
AM295005.1 | - | 8 - 9 | 997 | 127 | |
BC033674.1 | - | 8 - 9 | 862 | 127 |