The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105528.1 | NP_001098998.1 | 17 - 18 | 2071 | 74 |
NM_001308126.1 | NP_001295055.1 | 17 - 18 | 2071 | 74 | |
XM_011525948.1 | XP_011524250.1 | 18 - 19 | 2140 | 74 | |
XM_011525951.1 | XP_011524253.1 | 18 - 19 | 2140 | 74 | |
XM_017025721.1 | XP_016881210.1 | 18 - 19 | 2140 | 74 | |
XM_017025722.1 | XP_016881211.1 | 18 - 19 | 2161 | 74 | |
XM_017025723.1 | XP_016881212.1 | 18 - 19 | 2170 | 74 | |
XM_017025724.1 | XP_016881213.1 | 12 - 13 | 1704 | 74 | |
XM_017025725.1 | XP_016881214.1 | 12 - 13 | 2321 | 74 | |
GenBank mRNA | AK129955.1 | - | 5 - 6 | 570 | 74 |
BC091517.1 | - | 18 - 19 | 2112 | 74 | |
BC130535.1 | - | 17 - 18 | 2008 | 74 | |
BC144178.1 | - | 17 - 18 | 2008 | 74 |