The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105528.1 | NP_001098998.1 | 21 - 22 | 2664 | 110 |
NM_001308126.1 | NP_001295055.1 | 22 - 23 | 2736 | 110 | |
NM_198995.2 | NP_945346.2 | 21 - 22 | 2619 | 110 | |
XM_011525948.1 | XP_011524250.1 | 22 - 23 | 2733 | 110 | |
XM_011525951.1 | XP_011524253.1 | 21 - 22 | 2583 | 110 | |
XM_017025721.1 | XP_016881210.1 | 23 - 24 | 2805 | 110 | |
XM_017025722.1 | XP_016881211.1 | 23 - 24 | 2826 | 110 | |
XM_017025723.1 | XP_016881212.1 | 23 - 24 | 2835 | 110 | |
XM_017025724.1 | XP_016881213.1 | 17 - 18 | 2369 | 110 | |
XM_017025725.1 | XP_016881214.1 | 17 - 18 | 2986 | 110 | |
GenBank mRNA | AK000027.1 | - | 2 - 3 | 185 | 110 |
AK126038.1 | - | 21 - 22 | 2619 | 110 | |
AK129955.1 | - | 8 - 9 | 1013 | 110 | |
BC130535.1 | - | 21 - 22 | 2601 | 110 | |
BC144178.1 | - | 22 - 23 | 2673 | 110 |