The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018997.3 | NP_061870.1 | 1 - 2 | 607 | 168 |
GenBank mRNA | BC006554.2 | - | 1 - 2 | 512 | 168 |
BI755132.1 | - | 1 - 2 | 564 | 168 | |
U79258.1 | - | 1 - 2 | 528 | 168 |