The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000883.3 | NP_000874.2 | 15 - 16 | 1775 | 169 |
NM_001102605.1 | NP_001096075.1 | 14 - 15 | 1745 | 169 | |
NM_001142573.1 | NP_001136045.1 | 12 - 13 | 1545 | 169 | |
NM_001142574.1 | NP_001136046.1 | 12 - 13 | 1530 | 169 | |
NM_001142575.1 | NP_001136047.1 | 11 - 12 | 1470 | 169 | |
NM_001142576.1 | NP_001136048.1 | 14 - 15 | 1676 | 169 | |
NM_001304521.1 | NP_001291450.1 | 12 - 13 | 1573 | 169 | |
NM_183243.2 | NP_899066.1 | 13 - 14 | 1667 | 169 | |
XM_017012172.1 | XP_016867661.1 | 14 - 15 | 1863 | 169 | |
GenBank mRNA | AK054640.1 | - | 11 - 12 | 1442 | 169 |
AK054667.1 | - | 13 - 14 | 1667 | 169 | |
AK092452.1 | - | 14 - 15 | 1712 | 169 | |
AK122994.1 | - | 14 - 15 | 1744 | 169 | |
AK131294.1 | - | 12 - 13 | 1492 | 169 | |
AK293413.1 | - | 12 - 13 | 1502 | 169 | |
BC033622.1 | - | 13 - 14 | 1658 | 169 | |
CD014008.1 | - | 13 - 14 | 1459 | 169 | |
CD014010.1 | - | 14 - 15 | 1581 | 186 | |
J05272.1 | - | 15 - 16 | 2039 | 169 |