Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGA3
Entrez Gene ID
23163
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Gene Aliases
-
Chromosome Location
Chr.17: 75225464 - 75262393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172703.2 NP_001166174.1 7 - 8 684 133
NM_001172704.2 NP_001166175.1 6 - 7 585 133
NM_001291641.1 NP_001278570.1 8 - 9 855 133
NM_001291642.1 NP_001278571.1 4 - 5 426 133
NM_014001.4 NP_054720.1 6 - 7 587 133
NM_138619.3 NP_619525.1 7 - 8 686 133
XM_011524563.2 XP_011522865.1 7 - 8 660 133
XM_017024385.1 XP_016879874.1 7 - 8 660 133
XM_017024386.1 XP_016879875.1 6 - 7 549 133
XM_017024387.1 XP_016879876.1 8 - 9 809 133
XM_017024388.1 XP_016879877.1 8 - 9 821 133
XM_017024389.1 XP_016879878.1 7 - 8 654 133
XM_017024390.1 XP_016879879.1 6 - 7 553 133
XM_017024391.1 XP_016879880.1 6 - 7 539 133
XM_017024392.1 XP_016879881.1 6 - 7 608 133
GenBank mRNA AF190864.1 - 6 - 7 521 133
AF219138.1 - 7 - 8 621 133
AF219139.1 - 6 - 7 522 133
AK293291.1 - 6 - 7 547 133
AK296843.1 - 4 - 5 359 133
AK301895.1 - 6 - 7 553 133
AK302278.1 - 7 - 8 654 133
AK302423.1 - 7 - 8 707 133
AK304692.1 - 5 - 6 412 133
AL833603.1 - 8 - 9 795 133
BC063285.1 - 7 - 8 648 133
BC070044.1 - 6 - 7 526 133
D63876.1 - 6 - 7 521 133

View Full Product Details