Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS28
Entrez Gene ID
51160
Gene Name
VPS28, ESCRT-I subunit
Gene Aliases
-
Chromosome Location
Chr.8: 144423601 - 144428563 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016208.3 NP_057292.1 4 - 5 215 132
NM_183057.2 NP_898880.1 4 - 5 215 132
XM_005272323.3 XP_005272380.1 4 - 5 207 132
XM_005272324.3 XP_005272381.1 4 - 5 207 132
GenBank mRNA AF182844.1 - 3 - 4 167 132
AF316887.1 - 3 - 4 149 132
AK026865.1 - 4 - 5 165 132
BC006485.1 - 3 - 4 151 132
BC019321.1 - 3 - 4 151 132
BC050713.2 - 4 - 5 163 132
KU178635.1 - 3 - 4 106 132
KU178636.1 - 3 - 4 106 132
R72904.1 - 3 - 4 97 132

View Full Product Details