The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 31 - 32 | 4483 | 130 |
NM_003170.4 | NP_003161.2 | 30 - 31 | 4381 | 130 | |
XM_017024978.1 | XP_016880467.1 | 32 - 33 | 4542 | 130 | |
XM_017024979.1 | XP_016880468.1 | 30 - 31 | 4619 | 130 | |
XM_017024980.1 | XP_016880469.1 | 29 - 30 | 4313 | 130 | |
GenBank mRNA | AL833607.1 | - | 31 - 32 | 4484 | 130 |
BC003692.1 | - | 2 - 3 | 373 | 130 | |
BC003696.1 | - | 15 - 16 | 2256 | 130 | |
BC017105.1 | - | 10 - 11 | 1524 | 130 | |
BC073963.1 | - | 3 - 4 | 680 | 130 | |
BC136522.1 | - | 30 - 31 | 4293 | 130 | |
BC136524.1 | - | 30 - 31 | 4293 | 130 | |
BC150268.1 | - | 30 - 31 | 4295 | 130 | |
D79984.1 | - | 30 - 31 | 4295 | 130 |