The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 33 - 34 | 4724 | 156 |
NM_003170.4 | NP_003161.2 | 32 - 33 | 4622 | 156 | |
XM_017024978.1 | XP_016880467.1 | 34 - 35 | 4783 | 156 | |
XM_017024979.1 | XP_016880468.1 | 32 - 33 | 4860 | 156 | |
XM_017024980.1 | XP_016880469.1 | 31 - 32 | 4554 | 156 | |
GenBank mRNA | AL833607.1 | - | 33 - 34 | 4725 | 156 |
BC003692.1 | - | 4 - 5 | 614 | 156 | |
BC003696.1 | - | 17 - 18 | 2497 | 156 | |
BC017105.1 | - | 12 - 13 | 1765 | 156 | |
BC073963.1 | - | 5 - 6 | 921 | 156 | |
BC136522.1 | - | 32 - 33 | 4534 | 156 | |
BC136524.1 | - | 32 - 33 | 4534 | 156 | |
BC150268.1 | - | 32 - 33 | 4536 | 156 | |
D79984.1 | - | 32 - 33 | 4536 | 156 |