The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001894.1 | NP_001001894.1 | 18 - 19 | 1683 | 111 |
NM_001320703.1 | NP_001307632.1 | 18 - 19 | 1889 | 111 | |
NM_001320704.1 | NP_001307633.1 | 18 - 19 | 1879 | 111 | |
NM_003316.3 | NP_003307.3 | 18 - 19 | 2974 | 111 | |
XM_005261048.1 | XP_005261105.1 | 19 - 20 | 1789 | 111 | |
XM_005261049.1 | XP_005261106.1 | 19 - 20 | 1712 | 111 | |
XM_005261050.4 | XP_005261107.1 | 19 - 20 | 4674 | 111 | |
XM_005261052.1 | XP_005261109.1 | 18 - 19 | 1664 | 111 | |
XM_005261054.3 | XP_005261111.1 | 18 - 19 | 4618 | 111 | |
XM_005261055.1 | XP_005261112.1 | 9 - 10 | 844 | 111 | |
XM_005261056.2 | XP_005261113.1 | 9 - 10 | 923 | 111 | |
XM_011529739.2 | XP_011528041.1 | 11 - 12 | 995 | 111 | |
XM_011529740.2 | XP_011528042.1 | 16 - 17 | 1503 | 111 | |
XM_011529742.2 | XP_011528044.2 | 19 - 20 | 1734 | 111 | |
XM_017028458.1 | XP_016883947.1 | 19 - 20 | 1728 | 111 | |
XM_017028459.1 | XP_016883948.1 | 19 - 20 | 1727 | 111 | |
XM_017028460.1 | XP_016883949.1 | 18 - 19 | 1678 | 111 | |
XM_017028461.1 | XP_016883950.1 | 19 - 20 | 1789 | 111 | |
XM_017028462.1 | XP_016883951.1 | 19 - 20 | 1712 | 111 | |
XM_017028463.1 | XP_016883952.1 | 18 - 19 | 1741 | 111 | |
XM_017028464.1 | XP_016883953.1 | 9 - 10 | 963 | 111 | |
XM_017028465.1 | XP_016883954.1 | 15 - 16 | 3074 | 111 | |
XM_017028466.1 | XP_016883955.1 | 9 - 10 | 939 | 111 | |
XM_017028467.1 | XP_016883956.1 | 9 - 10 | 844 | 111 | |
GenBank mRNA | AJ001866.1 | - | 15 - 16 | 1735 | 111 |
AK291992.1 | - | 18 - 19 | 1791 | 111 | |
AK297921.1 | - | 10 - 11 | 970 | 111 | |
AK299991.1 | - | 9 - 10 | 820 | 111 | |
BC047867.1 | - | 12 - 13 | 1075 | 111 | |
BC063033.1 | - | 18 - 19 | 1701 | 111 | |
BC092466.1 | - | 18 - 19 | 1686 | 111 | |
BC137345.1 | - | 18 - 19 | 2113 | 111 | |
BG029806.1 | - | 3 - 4 | 290 | 111 | |
CR936616.1 | - | 3 - 4 | 279 | 111 | |
D83077.1 | - | 18 - 19 | 2974 | 111 | |
D83327.1 | - | 15 - 16 | 1326 | 111 | |
D84294.1 | - | 17 - 18 | 3047 | 111 | |
D84295.1 | - | 16 - 17 | 2961 | 111 | |
D84296.1 | - | 8 - 9 | 1385 | 111 |