The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002641.3 | NP_002632.1 | 6 - 6 | 2498 | 120 |
NM_020473.3 | NP_065206.3 | 5 - 5 | 1721 | 120 | |
NR_033835.1 | 6 - 6 | 2124 | 120 | ||
NR_033836.1 | 6 - 6 | 1840 | 120 | ||
XM_011545539.2 | XP_011543841.1 | 5 - 5 | 3322 | 120 | |
XM_017029581.1 | XP_016885070.1 | 5 - 5 | 2445 | 120 | |
GenBank mRNA | AK098047.1 | - | 5 - 5 | 1934 | 120 |
BC038236.1 | - | 5 - 5 | 2455 | 120 | |
D11466.1 | - | 6 - 6 | 2467 | 120 |