Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POPDC2
Entrez Gene ID
64091
Gene Name
popeye domain containing 2
Gene Aliases
POP2
Chromosome Location
Chr.3: 119642052 - 119660590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK226022.1 - 2 - 2 154 154

View Full Product Details