Product Details

Assay Reference Genome
Location

Chr.17:7702851 on build GRCh38
Cytoband
17p13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55135
Gene Symbol
WRAP53
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
Location
Chr.17:7686071-7703502 on build GRCh38
Assay Gene Location
Overlaps Exon 9 - Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRAP53 NM_001143990.1 NP_001137462.1
NM_001143991.1 NP_001137463.1
NM_001143992.1 NP_001137464.1
NM_018081.2 NP_060551.2
XM_011523952.2 XP_011522254.1
AK001247.1 BAA91579.1
AK056669.1
AK092370.1
AK225273.1
AK225411.1
AY766322.1
BC002336.2 AAH02336.1
DQ431240.1
DQ431241.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv952119 Chr17:7700483 - 7709982 on Build GRCh38 Deletion WRAP53 EFNB3

View Full Product Details