Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP54
Entrez Gene ID
144535
Gene Name
cilia and flagella associated protein 54
Gene Aliases
C12orf55, C12orf63
Chromosome Location
Chr.12: 96489566 - 96875555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306084.1 NP_001293013.1 59 - 60 8175 72
XM_011539069.2 XP_011537371.1 60 - 61 8375 72
XM_011539070.2 XP_011537372.1 60 - 61 8375 72
XM_011539071.2 XP_011537373.1 60 - 61 8375 72
XM_011539072.2 XP_011537374.1 60 - 61 8375 72
XM_011539073.2 XP_011537375.1 60 - 61 8375 72
XM_011539074.2 XP_011537376.1 60 - 61 8375 72
XM_011539076.2 XP_011537378.1 60 - 61 8375 72
XM_011539077.2 XP_011537379.1 59 - 60 8525 72
XM_011539078.2 XP_011537380.1 60 - 61 8375 72
XM_011539079.1 XP_011537381.1 56 - 57 7621 72
XM_011539085.1 XP_011537387.1 45 - 46 6196 72
XM_017018866.1 XP_016874355.1 60 - 61 8375 72
XM_017018867.1 XP_016874356.1 49 - 50 6627 72
GenBank mRNA AK126100.1 - 26 - 27 3489 72
DV080725.1 - 4 - 5 363 72

View Full Product Details