The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 20 - 21 | 2436 | 157 |
XM_011540849.1 | XP_011539151.1 | 20 - 21 | 2519 | 157 | |
XM_011540850.2 | XP_011539152.1 | 20 - 21 | 2448 | 157 | |
XM_011540851.1 | XP_011539153.1 | 20 - 21 | 2403 | 157 | |
XM_011540852.2 | XP_011539154.1 | 20 - 21 | 2385 | 157 | |
XM_011540855.1 | XP_011539157.1 | 18 - 19 | 2241 | 157 | |
XM_011540857.1 | XP_011539159.1 | 18 - 19 | 2000 | 157 | |
XM_011540859.2 | XP_011539161.1 | 17 - 18 | 1969 | 157 | |
XM_017000491.1 | XP_016855980.1 | 20 - 21 | 2380 | 157 | |
XM_017000492.1 | XP_016855981.1 | 14 - 15 | 1483 | 157 | |
XM_017000493.1 | XP_016855982.1 | 8 - 9 | 687 | 157 | |
XM_017000494.1 | XP_016855983.1 | 20 - 21 | 2380 | 157 | |
GenBank mRNA | AB044788.1 | - | 6 - 7 | 553 | 157 |
AB058637.1 | - | 19 - 20 | 2379 | 157 | |
AB204867.1 | - | 20 - 21 | 2437 | 157 | |
AK094079.1 | - | 3 - 4 | 263 | 157 | |
AK294754.1 | - | 14 - 15 | 1453 | 157 | |
AK302582.1 | - | 16 - 17 | 1604 | 157 | |
BC130525.1 | - | 5 - 6 | 382 | 157 | |
BC144172.1 | - | 5 - 6 | 382 | 157 |