Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP10-8
Entrez Gene ID
386681
Gene Name
keratin associated protein 10-8
Gene Aliases
KAP10.8, KRTAP18-8, KRTAP18.8
Chromosome Location
Chr.21: 44612079 - 44612954 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198695.2 NP_941968.2 1 - 1 36 62
GenBank mRNA AB076355.1 - 1 - 1 36 62
BC127016.1 - 1 - 1 30 62

View Full Product Details