The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198944.1 | NP_945182.1 | 1 - 1 | 49 | 80 |
XM_005259853.3 | XP_005259910.1 | 3 - 3 | 553 | 80 | |
XM_011527915.2 | XP_011526217.1 | 5 - 5 | 974 | 80 | |
GenBank mRNA | BC117212.1 | - | 1 - 1 | 124 | 80 |