The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021068.2 | NP_066546.1 | 1 - 1 | 775 | 106 |
GenBank mRNA | M27318.1 | - | 1 - 1 | 847 | 105 |