Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SULT1A1
- Entrez Gene ID
-
6817
- Gene Name
- sulfotransferase family 1A member 1
- Gene Aliases
-
HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
- Chromosome Location
-
Chr.16: 28605587 - 28623586 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177529.2 |
NP_803565.1 |
1 - 2 |
117 |
144 |
View Full Product Details