The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016044.2 | NP_057128.2 | 1 - 1 | 51 | 145 |
XM_011511283.2 | XP_011509585.1 | 1 - 1 | 51 | 145 | |
XM_011511284.2 | XP_011509586.1 | 1 - 1 | 60 | 145 | |
XM_011511285.2 | XP_011509587.1 | 1 - 1 | 51 | 145 | |
XM_011511286.2 | XP_011509588.1 | 1 - 1 | 60 | 145 | |
XM_011511287.2 | XP_011509589.1 | 1 - 1 | 51 | 145 | |
XM_017004236.1 | XP_016859725.1 | 1 - 1 | 51 | 145 | |
XM_017004237.1 | XP_016859726.1 | 1 - 1 | 60 | 145 | |
XM_017004238.1 | XP_016859727.1 | 1 - 1 | 51 | 145 | |
XM_017004239.1 | XP_016859728.1 | 1 - 1 | 60 | 145 | |
XM_017004240.1 | XP_016859729.1 | 1 - 1 | 51 | 145 | |
XM_017004242.1 | XP_016859731.1 | 1 - 1 | 60 | 145 | |
XM_017004244.1 | XP_016859733.1 | 1 - 1 | 60 | 145 | |
XM_017004247.1 | XP_016859736.1 | 1 - 1 | 60 | 145 | |
XM_017004248.1 | XP_016859737.1 | 1 - 1 | 51 | 145 | |
XM_017004249.1 | XP_016859738.1 | 1 - 1 | 51 | 145 | |
XM_017004250.1 | XP_016859739.1 | 1 - 1 | 51 | 145 | |
GenBank mRNA | AA311902.1 | - | 1 - 1 | 51 | 145 |
AF151863.1 | - | 1 - 1 | 51 | 144 |