The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033400.1 | 3 - 3 | 1103 | 141 | |
GenBank mRNA | BC029585.1 | - | 4 - 4 | 1047 | 141 |