Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPE
Entrez Gene ID
11316
Gene Name
coatomer protein complex subunit epsilon
Gene Aliases
epsilon-COP
Chromosome Location
Chr.19: 18899514 - 18919464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007263.3 NP_009194.2 10 - 10 1085 147
NM_199442.1 NP_955474.1 9 - 9 932 147
NM_199444.1 NP_955476.1 9 - 9 929 147
XM_011527655.1 XP_011525957.1 11 - 11 1228 147
XM_011527656.1 XP_011525958.1 10 - 10 1072 147
GenBank mRNA AF092438.1 - 10 - 10 1078 145
AI680336.1 - 1 - 1 147
AK222584.1 - 10 - 10 1082 147
AL136928.1 - 10 - 10 1062 147
BC007250.1 - 10 - 10 1062 147

View Full Product Details