Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN12
Entrez Gene ID
286256
Gene Name
lipocalin 12
Gene Aliases
-
Chromosome Location
Chr.9: 136949180 - 136955505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017014632.1 XP_016870121.1 3 - 4 519 74
GenBank mRNA AI681012.1 - 3 - 4 76

View Full Product Details