The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017001071.1 | XP_016856560.1 | 1 - 2 | 91 | 98 |
XM_017001073.1 | XP_016856562.1 | 1 - 2 | 91 | 98 | |
GenBank mRNA | AK302731.1 | - | 1 - 2 | 301 | 98 |