Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C9
Entrez Gene ID
1559
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
Chromosome Location
Chr.10: 94905867 - 94989395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000771.3 NP_000762.2 1 - 2 193 104
XM_017015758.1 XP_016871247.1 2 - 3 613 104
GenBank mRNA AK289420.1 - 1 - 2 193 104
AK298458.1 - 1 - 2 193 104
BC020754.1 - 1 - 2 194 104
BC070317.1 - 1 - 2 190 104
BC125054.1 - 1 - 2 168 104
BJ993098.1 - 1 - 2 193 104
D00173.1 - 1 - 2 159 104
M15331.1 - 1 - 2 153 104
M21939.1 - 1 - 2 154 104
M61855.1 - 1 - 2 180 104
M61857.1 - 1 - 2 178 104
S46963.1 - 1 - 2 131 104

View Full Product Details