The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175041.1 | NP_778206.1 | 6 - 7 | 755 | 173 |
GenBank mRNA | AB046400.1 | - | 5 - 6 | 623 | 173 |