The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001638.2 | NP_001629.1 | 2 - 2 | 1117 | 154 |
GenBank mRNA | BC026257.1 | - | 2 - 2 | 1117 | 154 |
CR936663.1 | - | 2 - 2 | 1142 | 154 | |
L27050.1 | - | 2 - 2 | 1112 | 153 |