Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPR88
Entrez Gene ID
54112
Gene Name
G protein-coupled receptor 88
Gene Aliases
COCPMR, STRG
Chromosome Location
Chr.1: 100538172 - 100542027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022049.2 NP_071332.2 2 - 2 3311 101
GenBank mRNA AB042410.1 - 2 - 2 3342 101
AF086060.1 - 1 - 1 498 101

View Full Product Details