The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272012.1 | NP_001258941.1 | 2 - 2 | 3928 | 99 |
NM_001272013.1 | NP_001258942.1 | 2 - 2 | 4048 | 99 | |
NM_033397.3 | NP_203755.1 | 3 - 3 | 4240 | 99 | |
XM_005270257.2 | XP_005270314.1 | 1 - 1 | 3804 | 99 | |
GenBank mRNA | AB051541.1 | - | 2 - 2 | 3959 | 99 |
AK055608.1 | - | 1 - 1 | 2651 | 99 | |
AK056526.1 | - | 2 - 2 | 3868 | 99 | |
AK094802.1 | - | 1 - 1 | 3280 | 99 | |
AY189283.1 | - | 1 - 1 | 2144 | 99 | |
BC040536.1 | - | 2 - 2 | 3900 | 99 | |
BC070108.1 | - | 3 - 3 | 4240 | 99 | |
DB114198.1 | - | 1 - 1 | 213 | 99 |