The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171020.1 | NP_001164491.1 | 4 - 4 | 4077 | 106 |
NM_152423.4 | NP_689636.3 | 5 - 5 | 4213 | 106 | |
XM_005262077.2 | XP_005262134.1 | 4 - 4 | 3994 | 106 | |
XM_011530856.1 | XP_011529158.1 | 4 - 4 | 4071 | 106 | |
XM_011530857.1 | XP_011529159.1 | 4 - 4 | 4040 | 106 | |
XM_017029269.1 | XP_016884758.1 | 5 - 5 | 4207 | 106 | |
GenBank mRNA | AA412505.1 | - | 1 - 1 | 106 | |
AK056478.1 | - | 3 - 3 | 3863 | 106 | |
AK090835.1 | - | 1 - 1 | 2729 | 106 |