Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPT1
Entrez Gene ID
85465
Gene Name
ethanolaminephosphotransferase 1
Gene Aliases
SELI, SEPI
Chromosome Location
Chr.2: 26346086 - 26395891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB051511.1 - 10 - 10 7888 148
AK091340.1 - 1 - 1 2240 148

View Full Product Details