Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC012036.1 - 2 703 4517792 Chr. 18 - 24044175 - 24045283 [+] on Build GRCh38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream TTC39C antisense RNA 1 TTC39C-AS1 -28.8kb Anti-sense Chr. - 23994213 - 24015460 on Build GRCh38
Downstream calcium binding tyrosine phosphorylation regulated CABYR CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2 93.7kb Sense Chr. - 24138956 - 24161600 on Build GRCh38

View Full Product Details