Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAIF1
Entrez Gene ID
203245
Gene Name
nuclear apoptosis inducing factor 1
Gene Aliases
C9orf90, bA379C10.2
Chromosome Location
Chr.9: 128061233 - 128067321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC021580.1 - 1 - 1 971 160

View Full Product Details