The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195643.1 | NP_001182572.1 | 2 - 2 | 3363 | 159 |
NM_176815.4 | NP_789785.1 | 2 - 2 | 3250 | 159 | |
XM_011512537.2 | XP_011510839.1 | 2 - 2 | 3428 | 159 | |
GenBank mRNA | AL832912.1 | - | 2 - 2 | 3039 | 159 |
BC045541.1 | - | 2 - 2 | 3081 | 159 |