The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_015379.3 | 3 - 3 | 879 | 106 | ||
GenBank mRNA | AK024303.1 | - | 2 - 2 | 1059 | 106 | |
AK092401.1 | - | 3 - 3 | 865 | 106 | ||
BC005351.1 | - | 3 - 3 | 731 | 106 | ||
BC017567.1 | - | 3 - 3 | 735 | 107 | ||
BC107722.1 | - | 3 - 3 | 735 | 106 | ||
DQ249310.1 | - | 3 - 3 | 827 | 106 | ||
DQ343132.1 | - | LIT3336 UCA1 |
3 - 3 | 879 | 106 | |
EU334869.1 | - | 3 - 3 | 880 | 106 | ||
GU357550.1 | - | 3 - 3 | 731 | 106 | ||
GU799561.1 | - | 3 - 3 | 879 | 106 | ||
GU799562.1 | - | 3 - 3 | 879 | 106 | ||
GU799563.1 | - | 3 - 3 | 879 | 106 | ||
GU799564.1 | - | 3 - 3 | 879 | 106 | ||
GU799565.1 | - | 3 - 3 | 879 | 106 | ||
HQ833208.1 | - | 3 - 3 | 879 | 106 | ||
HQ833209.1 | - | 3 - 3 | 879 | 106 |