The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002819.3 | 1 - 1 | 7781 | 100 | |
GenBank mRNA | EF177381.1 | - | 1 - 1 | 7781 | 100 |
FJ209305.1 | - | 1 - 1 | 6498 | 100 |