Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
Chromosome Location
Chr.9: 76441666 - 76507416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097633.1 NP_001091102.1 3 - 3 5340 150
NM_001097634.1 NP_001091103.1 3 - 3 5836 150
NM_001097635.1 NP_001091104.1 3 - 3 5376 150
NM_001097636.1 NP_001091105.1 2 - 2 5469 150
NM_001490.4 NP_001481.2 4 - 4 5458 150
XM_017014620.1 XP_016870109.1 5 - 5 6345 150
XM_017014621.1 XP_016870110.1 5 - 5 5482 150
GenBank mRNA AL832647.2 - 4 - 4 5377 150

View Full Product Details