The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197958.2 | NP_932062.1 | 2 - 2 | 554 | 102 |
GenBank mRNA | BC039153.1 | - | 2 - 2 | 410 | 102 |