Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEM1
Entrez Gene ID
374768
Gene Name
spermatid maturation 1
Gene Aliases
C17orf83
Chromosome Location
Chr.17: 7420360 - 7421632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK097400.1 - 2 - 2 1542 110

View Full Product Details