Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXorf21
Entrez Gene ID
80231
Gene Name
chromosome X open reading frame 21
Gene Aliases
-
Chromosome Location
Chr.X: 30558824 - 30577916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025159.2 NP_079435.1 3 - 3 1379 126
GenBank mRNA AK021639.1 - 3 - 3 1260 126
BC020611.1 - 3 - 3 1307 126

View Full Product Details