The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207312.2 | NP_997195.1 | 5 - 5 | 1452 | 141 |
GenBank mRNA | AK131043.1 | - | 5 - 5 | 1430 | 141 |
BC057811.1 | - | 5 - 5 | 1452 | 141 |