Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10K2
Entrez Gene ID
391107
Gene Name
olfactory receptor family 10 subfamily K member 2
Gene Aliases
OR1-4
Chromosome Location
Chr.1: 158419928 - 158420866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004476.1 NP_001004476.1 1 - 1 624 103

View Full Product Details