The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032409.2 | NP_115785.1 | 2 - 2 | 546 | 72 |
GenBank mRNA | AB053323.1 | - | 2 - 2 | 546 | 72 |
AF316873.1 | - | 2 - 2 | 522 | 72 | |
BC028215.1 | - | 2 - 2 | 461 | 72 |