The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003212.3 | NP_003203.1 | 2 - 3 | 472 | 102 |
GenBank mRNA | AK313319.1 | - | 2 - 3 | 208 | 102 |
AU125451.1 | - | 1 - 1 | 407 | 102 | |
BC022393.1 | - | 2 - 3 | 270 | 102 | |
BC067844.1 | - | 2 - 3 | 270 | 102 | |
DA941183.1 | - | 2 - 3 | 472 | 102 | |
X14253.1 | - | 2 - 3 | 335 | 102 |