The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005416.1 | NP_001005416.1 | 3 - 4 | 540 | 150 |
XM_006722763.3 | XP_006722826.1 | 3 - 4 | 483 | 150 |